
C₁₈-Reversed Phase Silica Gel Chromatography Media

Synonym(s):
Reversed phase silica gel 90 C18, Reversed phase silica gel C18
EC Number:
Products
Product Category
Brand
Physical Form
Separation Technique
Quality Segment
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-2 of 2