Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
universal fit pipet tips
Products
Product Category
pipette tips (15)
Manufacturer
Volume Range (µL)
Color
Package Size
Special Grade
Natural (7)
Sterility
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
CLS4860
Corning
®
universal fit pipet tips,
1-200 μL universal fit racked pipet tips, yellow, sterile, 10 racks/case, 960 tips/case
Expand
View Pricing
CLS4867
Corning
®
universal fit pipet tips,
100-1000 μL universal fit racked pipet tips, blue, non-sterile, 10 racks/case, 1000 tips/case
Expand
View Pricing
CLS4863
Corning
®
universal fit pipet tips,
1-200 μL universal fit racked pipet tips, natural, non-sterile, 10 racks/case, 960 tips/case
Expand
View Pricing
CLS4868
Corning
®
universal fit pipet tips,
100-1000 μL universal fit bulk packed pipet tips, blue, non-sterile, 1000 tips/bag, 1000 tips/case
Expand
View Pricing
CLS9032
Corning
®
universal fit pipet tips,
100-1000 μL universal fit racked pipet tips, blue, sterile, 10 racks/case, 1000 tips/case
Expand
View Pricing
CLS4806
Corning
®
universal fit pipet tips,
1-200 μL universal fit stack rack pipet tips, natural, non-sterile, 10 racks/case, 4800 tips/case
Expand
View Pricing
CLS4866
Corning
®
universal fit pipet tips,
1-200 μL universal fit bulk packed pipet tips, yellow, non-sterile, 1000 tips/bag, 1000 tips/case
Expand
View Pricing
CLS4864
Corning
®
universal fit pipet tips,
1-200 μL universal fit racked pipet tips, natural, sterile, 10 racks/case, 960 tips/case
Expand
View Pricing
CLS4862
Corning
®
universal fit pipet tips,
1-200 μL universal fit bulk packed pipet tips, natural, non-sterile, 1000 tips/bag, 1000 tips/case
Expand
View Pricing
CLS4804
Corning
®
universal fit pipet tips,
1-200 μL universal fit stack rack pipet tips, natural, sterile, 10 racks/case, 4800 tips/case
Expand
View Pricing
CLS4846
Corning
®
universal fit pipet tips,
1-1000 μL universal fit bulk packed pipet tips, blue, non-sterile, 1000 tips/bag, 10,000 tips/case
Expand
View Pricing
CLS4865
Corning
®
universal fit pipet tips,
1-200 μL universal fit racked pipet tips, yellow, non-sterile, 10 racks/case, 960 tips/case
Expand
View Pricing
CLS4845
Corning
®
universal fit pipet tips,
1-200 μL universal fit bulk packed pipet tips, yellow, non-sterile, 1000 tips/bag, 10,000 tips/case
Expand
View Pricing
CLS4844
Corning
®
universal fit pipet tips,
1-200 μL universal fit bulk packed pipet tips, natural, non-sterile, 1000 tips/bag, 10,000 tips/case
Expand
View Pricing
CLS4803
Corning
®
universal fit pipet tips,
1-200 μL universal fit stack rack pipet tips, natural, non-sterile, 10 racks/case, 4800 tips/case
Expand
View Pricing