Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
NBS
™
384 well microplates
Products
Product Category
plates (6)
Color
Manufacturer
Binding Type
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS3575
Corning
®
NBS
™
384 well microplates,
NBS
™
(non-binding surface), black polystyrene, flat bottom, non-sterile, lid: no, pack of 50
Expand
View Pricing
CLS3820
Corning
®
NBS
™
384 well microplates,
surface treatment, Non-binding Surface, black polystyrene, flat bottom, non-sterile, lid: no, pack of 10
Expand
View Pricing
CLS3574
Corning
®
NBS
™
384 well microplates,
surface treatment, Non-binding Surface, white polystyrene, flat bottom, non-sterile, lid: no, pack of 10
Expand
View Pricing
CLS3575BC
Corning
®
NBS
™
384 well microplates,
NBS
™
(non-binding surface), black polystyrene, flat bottom, non-sterile, with generic barcode, lid: no, pack of 10
Expand
View Pricing
CLS3640
Corning
®
NBS
™
384 well microplates,
surface treatment, Non-binding Surface, clear polystyrene, flat bottom, non-sterile, lid: no, pack of 25
Expand
View Pricing
CLS3574BC
Corning
®
NBS
™
384 well microplates,
NBS
™
(non-binding surface), white polystyrene, flat bottom, non-sterile, with generic barcode, lid: no, pack of 10
Expand
View Pricing