Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
narrow mouth reagent bottle, reusable
Products
Product Category
bottles (4)
Manufacturer
Capacity (L)
Capacity (mL)
OD (mm)
Cap
Color
Vial Bottle Color
Package Size
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS1500P250
Corning
®
narrow mouth reagent bottle, reusable,
capacity 250 mL, polypropylene, with GL-45 PP screw cap
Expand
View Pricing
CLS1500P2L
Corning
®
narrow mouth reagent bottle, reusable,
capacity 2 L, polypropylene, with GL-63 PP screw cap
Expand
View Pricing
CLS1500P500
Corning
®
narrow mouth reagent bottle, reusable,
capacity 500 mL, polypropylene, with GL-45 PP screw cap
Expand
View Pricing
CLS1500P1L
Corning
®
narrow mouth reagent bottle, reusable,
capacity 1 L, polypropylene, with GL-63 PP screw cap
Expand
View Pricing