Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
microcarriers
Products
Manufacturer
Physical Form
Package Size
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
CLS3781
Corning
®
microcarriers,
low concentration Synthemax
®
II, weight 10 g
Expand
View Pricing
CLS3786
Corning
®
microcarriers,
collagen coated, weight 10 g
Expand
View Pricing
CLS3779
Corning
®
microcarriers,
enhanced attachment, weight 10 g
Expand
View Pricing
CLS4622
Corning
®
microcarriers,
low concentration synthemax II, weight 100 g
Expand
View Pricing
CLS4620
Corning
®
microcarriers,
enhanced attachment, weight 100 g
Expand
View Pricing
CLS3772
Corning
®
microcarriers,
non-treated
, weight 10 g
Expand
View Pricing
CLS4623
Corning
®
microcarriers,
low concentration synthemax II, weight 500 g
Expand
View Pricing
CLS4621
Corning
®
microcarriers,
enhanced attachment, weight 500 g
Expand
View Pricing
CLS4625
Corning
®
microcarriers,
untreated, weight 500 g
Expand
View Pricing
CLS4624
Corning
®
microcarriers,
untreated, weight 100 g
Expand
View Pricing