Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Ice Pan, rectangular with lid
Products
Color
Manufacturer
Length (cm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
CLS432100
Corning
®
Ice Pan, rectangular with lid,
maxi, capacity 9L, blue
Expand
View Pricing
CLS432110
Corning
®
Ice Pan, rectangular with lid,
midi, capacity 4L, blue
Expand
View Pricing
CLS432112
Corning
®
Ice Pan, rectangular with lid,
midi, capacity 4L, lime green
Expand
View Pricing
CLS432102
Corning
®
Ice Pan, rectangular with lid,
maxi, capacity 9L, purple
Expand
View Pricing
CLS432101
Corning
®
Ice Pan, rectangular with lid,
maxi, capacity 9L, lime green
Expand
View Pricing
CLS432113
Corning
®
Ice Pan, rectangular with lid,
midi, capacity 4L, pink
Expand
View Pricing
CLS432114
Corning
®
Ice Pan, rectangular with lid,
midi, capacity 4L, purple
Expand
View Pricing
CLS432111
Corning
®
Ice Pan, rectangular with lid,
midi, capacity 4L, orange
Expand
View Pricing