Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
hotplate and stirrer with digital display
Products
Product Category
heaters (5)
Color
Manufacturer
Length (in)
Package Size
Sterility
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
CLS6798420D
Corning
®
hotplate and stirrer with digital display,
AC/DC input 230 V AC (CEE7-7 plug), plate L × W 5 in. × 7 in.
Expand
View Pricing
CLS6795420D
Corning
®
hotplate and stirrer with digital display,
AC/DC input 120 V AC, US 3-pin plug, plate L × W 5 in. × 7 in.
Expand
View Pricing
CLS6795620D
Corning
®
hotplate and stirrer with digital display,
AC/DC input 120 V AC, US 3-pin plug, plate L × W 10 in. × 10 in.
Expand
View Pricing
CLS6796420D
Corning
®
hotplate and stirrer with digital display,
AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 2-pin Euro plug), plate L × W 5 in. × 7 in., CE compliant
Expand
View Pricing
CLS6796620D
Corning
®
hotplate and stirrer with digital display,
AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 2-pin Euro plug), plate L × W 10 in. × 10 in., CE compliant
Expand
View Pricing