Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Falcon
®
Round Bottom Test Tubes
Products
Product Category
tubes (14)
Manufacturer
Capacity (mL)
Cap
Package Size
Sterility
Filter Products
Showing 1-14 of 14
Compare
Product Number
Description
Pricing
CLS352235
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 5 mL, cap, cell-strainer, sterile, case of 500
Expand
View Pricing
CLS352054
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 5 mL, cap, snap, sterile, case of 1000
Expand
View Pricing
CLS352008
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 5 mL, cap: no, non-sterile, case of 1000
Expand
View Pricing
CLS352052
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 5 mL, cap: no, sterile, case of 500
Expand
View Pricing
CLS352058
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 5 mL, cap, snap, sterile, case of 500
Expand
View Pricing
CLS352057
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 14 mL, cap: no, sterile, case of 500
Expand
View Pricing
CLS352051
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 14 mL, cap, snap, sterile, case of 1000
Expand
View Pricing
CLS352003
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 5 mL, cap, snap, sterile, individually wrapped, case of 500
Expand
View Pricing
CLS352027
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 8 mL, cap, screw, sterile
Expand
View Pricing
CLS352025
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 16 mL, cap, screw, sterile, case of 1000
Expand
View Pricing
CLS352037
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 16 mL, cap, screw, sterile, individually wrapped, case of 500
Expand
View Pricing
CLS352017
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 14 mL, cap: no, sterile, case of 1000
Expand
View Pricing
CLS352045
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 19 mL, cap, screw, sterile, individually wrapped, case of 500
Expand
View Pricing
CLS352001
Corning
®
Falcon
®
Round Bottom Test Tubes,
capacity 14 mL, cap, snap, sterile, individually wrapped, case of 500
Expand
View Pricing