Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
Costar
®
Stripette
®
serological pipettes, individually plastic wrapped
Products
Product Category
pipettes (9)
Color
Manufacturer
Capacity (mL)
ID (mm)
Length (mm)
OD (mm)
Package Size
Sterility
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
CLS4101
Corning
®
Costar
®
Stripette
®
serological pipettes, individually plastic wrapped,
capacity 10 mL, case of 200
Expand
View Pricing
CLS4251
Corning
®
Costar
®
Stripette
®
serological pipettes, individually plastic wrapped,
capacity 25 mL
Expand
View Pricing
CLS4051
Corning
®
Costar
®
Stripette
®
serological pipettes, individually plastic wrapped,
capacity 5 mL, case of 200
Expand
View Pricing
CLS4501
Corning
®
Costar
®
Stripette
®
serological pipettes, individually plastic wrapped,
capacity 50 mL, case of 100
Expand
View Pricing
CLS4484
Corning
®
Costar
®
Stripette
®
serological pipettes, individually plastic wrapped,
capacity 100 mL, case of 100
Expand
View Pricing
CLS4021
Corning
®
Costar
®
Stripette
®
serological pipettes, individually plastic wrapped,
capacity 2 mL, case of 1000
Expand
View Pricing
CLS4492
Corning
®
Costar
®
Stripette
®
serological pipettes, individually plastic wrapped,
capacity 10 mL, case of 200
Expand
View Pricing
CLS4011
Corning
®
Costar
®
Stripette
®
serological pipettes, individually plastic wrapped,
capacity 1 mL, case of 1000
Expand
View Pricing
CLS4012
Corning
®
Costar
®
Stripette
®
serological pipettes, individually plastic wrapped,
capacity 1 mL, case of 200
Expand
View Pricing