Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
CellSTACK
®
accessories
Products
Product Category
caps (3)
Color
Manufacturer
ID (mm)
Package Size
Sterility
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
CLS3968
Corning
®
CellSTACK
®
accessories,
vented cap
Expand
View Pricing
CLS3281
Corning
®
CellSTACK
®
accessories,
tubing I.D. 9.5 mm, L 7 cm, filter size 50 mm, PVDF membrane, sterile, vent cap, case of 5
Expand
View Pricing
CLS3282
Corning
®
CellSTACK
®
accessories,
I.D. 3.2 mm, sterile, female Luer Lock with male luer plug, cap (fill), case of 5
Expand
View Pricing
CLS3969
Corning
®
CellSTACK
®
accessories,
screw cap, not vented
Expand
View Pricing
CLS11902
Corning
®
CellSTACK
®
accessories,
filling accessory, C-Flex tubing diam.
3
/
8
in., female MPC connector, sterile
Expand
View Pricing
CLS3732
Corning
®
CellSTACK
®
accessories,
universal cap, diam. 33 mm, sterile
Expand
View Pricing
CLS3328
Corning
®
CellSTACK
®
accessories,
sterile, fill cap, female MPC coupling, 6.4 mm I.D. barbed fitting with male end cap, cap (fill)
Expand
View Pricing
CLS3284
Corning
®
CellSTACK
®
accessories,
tubing I.D. 9.5 mm, L 7 cm, filter size 37 mm , hydrophobic glass fiber, sterile, cap (vent), case of 4
Expand
View Pricing
CLS3329
Corning
®
CellSTACK
®
accessories,
sterile, female MPC coupling with male end cap, 9.5 mm I.D. barbed fitting with male end cap, cap (fill)
Expand
View Pricing
CLS3339
Corning
®
CellSTACK
®
accessories,
sterile, male MPC coupling with male end cap, 9.5 mm I.D. barbed fitting with female end cap, cap (fill), case of 4
Expand
View Pricing
CLS3331
Corning
®
CellSTACK
®
accessories,
stacking device, ABS
Expand
View Pricing
CLS3333
Corning
®
CellSTACK
®
accessories,
sterile, 6.4 mm I.D. tubing, 70 cm length, male MPC coupling with female end cap, cap (fill), case of 4
Expand
View Pricing
CLS3324
Corning
®
CellSTACK
®
accessories,
two vented overcaps (orange) and one solid overcap (red) for the universal cap
Expand
View Pricing
CLS3283
Corning
®
CellSTACK
®
accessories,
I.D. 9.5 mm, size 7.94 mm , barbed fitting, sterile, cap (fill), case of 5
Expand
View Pricing
CLS3334
Corning
®
CellSTACK
®
accessories,
sterile, male MPC coupling, with male end cap, 6.4 mm I.D. barbed fitting with female end cap, cap (fill), case of 4
Expand
View Pricing