Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
CellBIND
®
Surface CellSTACK
®
cell culture chambers
Products
Product Category
flasks (6)
Manufacturer
Binding Type
Surface Area cm²
Working Volume (ml)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
CLS3320
Corning
®
CellBIND
®
Surface CellSTACK
®
cell culture chambers,
CellBIND
®
surface, polystyrene, sterile, 10 Chamber with Vent Caps, graduations, high-density polyethylene cap, case of 6
Expand
View Pricing
CLS3310
Corning
®
CellBIND
®
Surface CellSTACK
®
cell culture chambers,
CellBIND
®
surface, polystyrene, sterile, 2 Chamber with Vent Caps, graduations, orange high-density polyethylene cap, case of 5
Expand
View Pricing
CLS3330
Corning
®
CellBIND
®
Surface CellSTACK
®
cell culture chambers,
CellBIND
®
surface, polystyrene, sterile, 1 Chamber with Vent Caps, graduations, orange high-density polyethylene cap, case of 8
Expand
View Pricing
CLS3311
Corning
®
CellBIND
®
Surface CellSTACK
®
cell culture chambers,
CellBIND
®
surface, polystyrene, sterile, 5 Chamber with Vent Caps, graduations, high-density polyethylene cap, case of 2
Expand
View Pricing
CLS3312
Corning
®
CellBIND
®
Surface CellSTACK
®
cell culture chambers,
CellBIND
®
surface, polystyrene, sterile, 10 Chamber with Vent Caps, graduations, orange high-density polyethylene cap, case of 2
Expand
View Pricing
CLS3321
Corning
®
CellBIND
®
Surface CellSTACK
®
cell culture chambers,
CellBIND
®
surface, polystyrene, sterile, 40 Chamber with Vent Caps, graduations, orange high-density polyethylene cap, case of 2
Expand
View Pricing