Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
CellBIND
®
384 well microplate
Products
Product Category
plates (3)
Color
Manufacturer
Binding Type
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
CLS3770
Corning
®
CellBIND
®
384 well microplate,
CellBIND
®
surface, black polystyrene, flat bottom, sterile, lid, pack of 20
Expand
View Pricing
CLS3770BC
Corning
®
CellBIND
®
384 well microplate,
CellBIND
®
surface, black polystyrene, flat bottom, sterile, with generic barcode, lid, pack of 20
Expand
View Pricing
CLS4579
Corning
®
CellBIND
®
384 well microplate,
surface treatment, CellBIND
®
, clear polystyrene, flat bottom, sterile, lid, pack of 100
Expand
View Pricing