Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
1536 well plate, with generic barcode
Products
Product Category
plates (10)
Color
Manufacturer
Binding Type
Wells
Color
Plate Material
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
CLS3727BC
Corning
®
1536 well plate, with generic barcode,
size 1536 wells, flat bottom white polystyrene wells, sterile
Expand
View Pricing
CLS3838BC
Corning
®
1536 well plate, with generic barcode,
size 1536 wells, flat bottom clear polystyrene wells, sterile, lid
Expand
View Pricing
CLS3724BC
Corning
®
1536 well plate, with generic barcode,
size 1536 wells, non-treated, 50/cs, flat bottom black polystyrene wells, non-sterile, lid: no
Expand
View Pricing
CLS3893BC
Corning
®
1536 well plate, with generic barcode,
size 1536 wells, flat bottom clear polystyrene wells, black plate
Expand
View Pricing
CLS3728BC
Corning
®
1536 well plate, with generic barcode,
size 1536 wells, flat bottom wells black polystyrene wells, NBS surface, 50/cs, non-sterile
Expand
View Pricing
CLS3726BC
Corning
®
1536 well plate, with generic barcode,
size 1536 wells, non-treated, 50/cs, flat bottom black polystyrene wells, non-sterile, lid
Expand
View Pricing
CLS3832BC
Corning
®
1536 well plate, with generic barcode,
size 1536 wells, flat bottom clear polystyrene wells, CellBIND
®
surface, sterile, lid
Expand
View Pricing
CLS3731BC
Corning
®
1536 well plate, with generic barcode,
size 1536 wells, cellBIND
®
, flat bottom white polystyrene wells, sterile, lid
Expand
View Pricing
CLS3729BC
Corning
®
1536 well plate, with generic barcode,
size 1536 wells, NBS surface, flat bottom white polystyrene wells
Expand
View Pricing
CLS3859BC
Corning
®
1536 well plate, with generic barcode,
black polystyrene plate, sterile, lid
Expand
View Pricing