Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
1536 well plates
Products
Product Category
plates (12)
Color
Manufacturer
Binding Type
Surface Area cm²
Volume (µL)
Wells
Color
Plate Material
Package Size
Sterility
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
CLS3727
Corning
®
1536 well plates,
flat bottom white polystyrene wells, NBS
™
(TC-treated), 50/cs, sterile, lid
Expand
View Pricing
CLS3728
Corning
®
1536 well plates,
flat bottom black polystyrene wells, NBS
™
(TC-treated), 50/cs, non-sterile, lid: no
Expand
View Pricing
CLS3893
Corning
®
1536 well plates,
flat bottom clear polystyrene wells, TC-treated, 50/cs, sterile, lid
Expand
View Pricing
CLS3724
Corning
®
1536 well plates,
flat bottom black polystyrene wells, non-treated, non-sterile, lid: no
Expand
View Pricing
CLS3936
Corning
®
1536 well plates,
non-treated (medium bind), 50/cs, round bottom black polystyrene wells, non-sterile, lid: no
Expand
View Pricing
CLS3725
Corning
®
1536 well plates,
flat bottom white polystyrene wells, non-treated (medium bind), 50/cs, non-sterile, lid: no
Expand
View Pricing
CLS3729
Corning
®
1536 well plates,
flat bottom white polystyrene wells, NBS
™
(TC-treated), 50/cs, non-sterile, lid: no
Expand
View Pricing
CLS3831
Corning
®
1536 well plates,
low base, non-treated, non-sterile
Expand
View Pricing
CLS3838
Corning
®
1536 well plates,
flat bottom clear polystyrene wells, low base, TC-treated, sterile, lid
Expand
View Pricing
CLS3832
Corning
®
1536 well plates,
low base, sterile
Expand
View Pricing
CLS3891
Corning
®
1536 well plates,
flat bottom clear polystyrene wells, non-treated, 50/cs, non-sterile, lid: no
Expand
View Pricing
CLS3937
Corning
®
1536 well plates,
round bottom white polystyrene wells, non-treated (medium bind), 50/cs, non-sterile, lid: no
Expand
View Pricing