Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper indium sulfide zinc sulfide (CuInS
2
/ZnS) core/shell Quantum Dots
Linear Formula:
Cu
InS
2
/ZnS
Products
Product Category
quantum dots (3)
Brand
Sigma-Aldrich (7)
Quality Segment
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
924342
Copper indium sulfide zinc sulfide (CuInS
2
/ZnS) core/shell Quantum Dots,
in toluene, fluorescence λ
em
750 nm, 1 mg/mL
Expand
View Pricing
924261
Copper indium sulfide zinc sulfide (CuInS
2
/ZnS) core/shell Quantum Dots,
in toluene, fluorescence λ
em
530 nm, 1 mg/mL
Expand
View Pricing
924326
Copper indium sulfide zinc sulfide (CuInS
2
/ZnS) core/shell Quantum Dots,
in toluene, fluorescence λ
em
650 nm, 1 mg/mL
Expand
View Pricing
924288
Copper indium sulfide zinc sulfide (CuInS
2
/ZnS) core/shell Quantum Dots,
in toluene, fluorescence λ
em
560 nm, 1 mg/mL
Expand
View Pricing
924334
Copper indium sulfide zinc sulfide (CuInS
2
/ZnS) core/shell Quantum Dots,
in toluene, fluorescence λ
em
700 nm, 1 mg/mL
Expand
View Pricing
924296
Copper indium sulfide zinc sulfide (CuInS
2
/ZnS) core/shell Quantum Dots,
in toluene, fluorescence λ
em
590 nm, 1 mg/mL
Expand
View Pricing
924318
Copper indium sulfide zinc sulfide (CuInS
2
/ZnS) core/shell Quantum Dots,
in toluene, fluorescence λ
em
620 nm, 1 mg/mL
Expand
View Pricing