Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper(II) sulfate
Synonym(s):
Cupric sulfate
Linear Formula:
CuSO
4
CAS Number:
7758-98-7
Molecular Weight:
159.61
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (5)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reaction Type
Special Grade
Puriss (2)
Puriss P.A. (1)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
C1297
Copper(II) sulfate,
ReagentPlus
®
, ≥99%
Expand
View Pricing
451657
Copper(II) sulfate,
anhydrous, powder, ≥99.99% trace metals basis
Expand
View Pricing
61230
Copper(II) sulfate,
puriss. p.a., anhydrous, ≥99.0% (RT)
Expand
View Pricing
12852
Copper(II) sulfate,
puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99-100.5% (based on anhydrous substance)
Expand
View Pricing
908940
Copper(II) sulfate,
free-flowing, Redi-Dri
™
,
ReagentPlus
®
, ≥99%
Expand
View Pricing