Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper(I) bromide
Synonym(s):
Cuprous bromide
Linear Formula:
CuBr
CAS Number:
7787-70-4
Molecular Weight:
143.45
EC Number:
232-131-6
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (5)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Reaction Type
Reagent Type
Core
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
212865
Copper(I) bromide,
98%
Expand
View Pricing
254185
Copper(I) bromide,
99.999% trace metals basis
Expand
View Pricing
923060
Copper(I) bromide,
99.9% trace metals basis
Expand
View Pricing
923052
Copper(I) bromide,
99.99% trace metals basis
Expand
View Pricing
735906
Copper(I) bromide,
AnhydroBeads
™
, -10 mesh, 99.99% trace metals basis
Expand
View Pricing