Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Copper-beryllium alloy, Cu98Be2
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
GF19575243
Copper-beryllium alloy, Cu98Be2,
foil, 0.5m coil, 0.2mm thickness, half hard
Expand
View Pricing
GF46504087
Copper-beryllium alloy, Cu98Be2,
2000m wire, 0.1mm diameter, hard
Expand
View Pricing
GF10811764
Copper-beryllium alloy, Cu98Be2,
foil, 25 x 25mm, 0.005mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF58597217
Copper-beryllium alloy, Cu98Be2,
2m wire, 0.5mm diameter, hard
Expand
View Pricing
GF20549583
Copper-beryllium alloy, Cu98Be2,
foil, 10mm disks, 0.0065mm thickness, as rolled
Expand
View Pricing
GF17485487
Copper-beryllium alloy, Cu98Be2,
1000mm wire, 1.0mm diameter, hard, straight wire
Expand
View Pricing
GF70636866
Copper-beryllium alloy, Cu98Be2,
foil, 100 x 100mm, 1.0mm thickness, annealed
Expand
View Pricing
GF17025612
Copper-beryllium alloy, Cu98Be2,
foil, 100 x 100mm, 0.2mm thickness, half hard
Expand
View Pricing
GF34028434
Copper-beryllium alloy, Cu98Be2,
foil, 1m coil, 0.05mm thickness, quarter hard
Expand
View Pricing
GF69508682
Copper-beryllium alloy, Cu98Be2,
50m wire, 0.30mm diameter, annealed
Expand
View Pricing
GF12043471
Copper-beryllium alloy, Cu98Be2,
foil, 100 x 100mm, 0.25mm thickness, half hard
Expand
View Pricing
GF07068045
Copper-beryllium alloy, Cu98Be2,
foil, 50 x 50mm, 0.2mm thickness, annealed
Expand
View Pricing
GF74714968
Copper-beryllium alloy, Cu98Be2,
foil, 50 x 50mm, 0.38mm thickness, annealed
Expand
View Pricing
GF65397718
Copper-beryllium alloy, Cu98Be2,
foil, 25 x 25mm, 0.38mm thickness, annealed
Expand
View Pricing
GF45829075
Copper-beryllium alloy, Cu98Be2,
foil, 25 x 25mm, 0.1mm thickness, half hard
Expand
View Pricing
GF66210218
Copper-beryllium alloy, Cu98Be2,
foil, 100 x 100mm, 0.02mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF75463511
Copper-beryllium alloy, Cu98Be2,
foil, 8mm disks, 0.01mm thickness, as rolled
Expand
View Pricing
GF69589692
Copper-beryllium alloy, Cu98Be2,
500m wire, 0.5mm diameter, hard
Expand
View Pricing
GF91137269
Copper-beryllium alloy, Cu98Be2,
250m wire, 0.1mm diameter, hard
Expand
View Pricing
GF60565091
Copper-beryllium alloy, Cu98Be2,
foil, 50 x 50mm, 1.0mm thickness, annealed
Expand
View Pricing
Page 1 of 11
Page 1 of 11