Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Constantan - resistance alloy, Cu55Ni45
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 196
Page 1 of 10
Page 1 of 10
Compare
Product Number
Description
Pricing
GF26506526
Constantan - resistance alloy, Cu55Ni45,
rod, 50mm x 25.4mm diameter, cold finished
Expand
View Pricing
GF25830121
Constantan - resistance alloy, Cu55Ni45,
rod, 500mm x 3.2mm diameter, annealed
Expand
View Pricing
GF52777963
Constantan - resistance alloy, Cu55Ni45,
100m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF45211957
Constantan - resistance alloy, Cu55Ni45,
50m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF29918144
Constantan - resistance alloy, Cu55Ni45,
foil, 0.5m coil, 150mm coil width, 0.025mm thickness, as rolled
Expand
View Pricing
GF13284222
Constantan - resistance alloy, Cu55Ni45,
100m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF34364493
Constantan - resistance alloy, Cu55Ni45,
10m wire, 0.025mm diameter, annealed
Expand
View Pricing
GF60049931
Constantan - resistance alloy, Cu55Ni45,
10m wire, 0.125mm diameter, annealed
Expand
View Pricing
GF50752885
Constantan - resistance alloy, Cu55Ni45,
insulated wire, 100m length, 0.1mm conductor diameter, 0.01mm insulation thickness, polyimide insulation
Expand
View Pricing
GF29632541
Constantan - resistance alloy, Cu55Ni45,
foil, 100 x 100mm, 0.05mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF06276517
Constantan - resistance alloy, Cu55Ni45,
insulated wire, 20m length, 0.07mm conductor diameter, 0.01mm insulation thickness, polyurethane insulation
Expand
View Pricing
GF83032703
Constantan - resistance alloy, Cu55Ni45,
10m wire, 0.05mm diameter, annealed
Expand
View Pricing
GF83743746
Constantan - resistance alloy, Cu55Ni45,
rod, 200mm x 8mm diameter, annealed
Expand
View Pricing
GF35370424
Constantan - resistance alloy, Cu55Ni45,
rod, 200mm x 3.2mm diameter, annealed
Expand
View Pricing
GF95954158
Constantan - resistance alloy, Cu55Ni45,
rod, 1000mm x 8mm diameter, annealed
Expand
View Pricing
GF00789497
Constantan - resistance alloy, Cu55Ni45,
50m wire, 0.5mm diameter, annealed
Expand
View Pricing
GF05706277
Constantan - resistance alloy, Cu55Ni45,
50m wire, 0.25mm diameter, annealed
Expand
View Pricing
GF05066951
Constantan - resistance alloy, Cu55Ni45,
foil, 100 x 100mm, 0.02mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF92147991
Constantan - resistance alloy, Cu55Ni45,
foil, 100 x 100mm, 0.0125mm thickness, as rolled, light tight (lt)
Expand
View Pricing
GF67967634
Constantan - resistance alloy, Cu55Ni45,
rod, 500mm x 8mm diameter, annealed
Expand
View Pricing
Page 1 of 10
Page 1 of 10