Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Collagenase from
Clostridium histolyticum
Synonym(s):
Clostridiopeptidase A
CAS Number:
9001-12-1
EC Number:
232-582-9
Products
Product Category
enzymes (20)
Brand
Sigma-Aldrich (23)
Biological Source
Physical Form
Quality Segment
Application
Sterilization Method
Technique
Filter Products
Showing 1-20 of 23
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
C5138
Collagenase from
Clostridium histolyticum
,
suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Expand
View Pricing
C0130
Collagenase from
Clostridium histolyticum
,
for general use, Type I, ≥125 CDU/mg solid
Expand
View Pricing
C6885
Collagenase from
Clostridium histolyticum
,
suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Expand
View Pricing
C2139
Collagenase from
Clostridium histolyticum
,
suitable for release of rat epididymal adipocytes and hepatocytes (for methodology see Type II and Type IV), Type VIII, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Expand
View Pricing
C9891
Collagenase from
Clostridium histolyticum
,
Type IA, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid, For general use
Expand
View Pricing
C7657
Collagenase from
Clostridium histolyticum
,
Type XI, 2-5 FALGPA units/mg solid, ≥800 CDU/mg solid
Expand
View Pricing
C9263
Collagenase from
Clostridium histolyticum
,
Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
Expand
View Pricing
C9407
Collagenase from
Clostridium histolyticum
,
powder, Suitable for the digestion and isolation of physiologically active pancreatic islet cells, suitable for cell culture
Expand
View Pricing
C2674
Collagenase from
Clostridium histolyticum
,
lyophilized powder, ≥125 CDU/mg solid (CDU = collagen digestion units), 0.5-5.0 FALGPA units/mg solid
Expand
View Pricing
C9722
Collagenase from
Clostridium histolyticum
,
lyophilized powder (from 0.2μm filtered solution), 0.5-5.0 FALGPA units/mg solid, suitable for cell culture
Expand
View Pricing
C2399
Collagenase from
Clostridium histolyticum
,
0.2 μm filtered, high purity, purified by chromatography, Type VII-S, ≥4 FALGPA units/mg solid, ≥700 CDU/mg solid (CDU = collagen digestion units)
Expand
View Pricing
C1889
Collagenase from
Clostridium histolyticum
,
0.2 μm filtered, suitable for release of physiologically active rat hepatocytes, Type IV-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Expand
View Pricing
C0773
Collagenase from
Clostridium histolyticum
,
high purity, purified by chromatography, Type VII, ≥4 FALGPA units/mg solid, lyophilized powder, ≥700 CDU/mg solid (CDU = collagen digestion units)
Expand
View Pricing
C9697
Collagenase from
Clostridium histolyticum
,
lyophilized powder (from 0.2 μm filtered solution), suitable for cell culture
Expand
View Pricing
C2799
Collagenase from
Clostridium histolyticum
,
powder, suitable for cell culture, ≥4 FALGPA units/mg solid, high purity, ≥700 CDU/mg solid (CDU = collagen digestion units)
Expand
View Pricing
C1639
Collagenase from
Clostridium histolyticum
,
0.2 μm filtered, for general use, Type I-S, 0.2-1.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Expand
View Pricing
C1764
Collagenase from
Clostridium histolyticum
,
0.2 μm filtered, suitable for release of physiologically active rat epididymal adipocytes, Type II-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Expand
View Pricing
C8051
Collagenase from
Clostridium histolyticum
,
Sigma Blend Type H, ≥1.0 FALGPA units/mg solid
Expand
View Pricing
C5894
Collagenase from
Clostridium histolyticum
,
non-sterile; 0.2 μm filtered, Type IA-S, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Expand
View Pricing
C7926
Collagenase from
Clostridium histolyticum
,
Sigma Blend Type F, ≥2.0 FALGPA units/mg solid
Expand
View Pricing
Page 1 of 2
Page 1 of 2