Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cochranes atom centers for Orbit molecular models
Products
Color
Manufacturer
Filter Products
Showing 1-20 of 27
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
Z175501
Cochranes atom centers for Orbit molecular models,
sulfur, octahedral, yellow
Expand
View Pricing
Z127175
Cochranes atom centers for Orbit molecular models,
carbon, tetrahedral, black
Expand
View Pricing
Z159298
Cochranes atom centers for Orbit molecular models,
carbon, trigonal bipyramid, black
Expand
View Pricing
Z127124
Cochranes atom centers for Orbit molecular models,
hydrogen, univalent, white
Expand
View Pricing
Z127140
Cochranes atom centers for Orbit molecular models,
oxygen, diunivalent/hydrogen bonding, red
Expand
View Pricing
Z159921
Cochranes atom centers for Orbit molecular models,
oxygen, trigonal, red
Expand
View Pricing
Z127183
Cochranes atom centers for Orbit molecular models,
nitrogen, tetrahedral, blue
Expand
View Pricing
Z159263
Cochranes atom centers for Orbit molecular models,
oxygen, tetrahedral, red
Expand
View Pricing
Z150983
Cochranes atom centers for Orbit molecular models,
carbon, octahedral, black
Expand
View Pricing
Z185515
Cochranes atom centers for Orbit molecular models,
halogen, octahedral, green
Expand
View Pricing
Z159573
Cochranes atom centers for Orbit molecular models,
hydrogen boding, red
Expand
View Pricing
Z127159
Cochranes atom centers for Orbit molecular models,
carbon, 5 ring planar, black
Expand
View Pricing
Z146188
Cochranes atom centers for Orbit molecular models,
nitrogen, amine,amide, blue
Expand
View Pricing
Z127167
Cochranes atom centers for Orbit molecular models,
carbon, 6 ring planar, black
Expand
View Pricing
Z185280
Cochranes atom centers for Orbit molecular models,
fluorine, univalent, light green
Expand
View Pricing
Z159883
Cochranes atom centers for Orbit molecular models,
phosphorus, trivalent, purple
Expand
View Pricing
Z185337
Cochranes atom centers for Orbit molecular models,
phosphorus, trigonal bipyramid, purple
Expand
View Pricing
Z148474
Cochranes atom centers for Orbit molecular models,
metal, 12 coordinate, silver
Expand
View Pricing
Z159824
Cochranes atom centers for Orbit molecular models,
bromine, univalent, blue-green
Expand
View Pricing
Z129577
Cochranes atom centers for Orbit molecular models,
nitrogen, 6 ring planar/azo, blue
Expand
View Pricing
Page 1 of 2
Page 1 of 2