Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Closures for Snap Ring vials
Products
Product Category
caps (8)
Brand
Supelco (7)
Diameter (mm)
ID (mm)
OD (mm)
Closure Type
Color
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
27427
Closures for Snap Ring vials,
polypropylene seal, PTFE/silicone (with starburst), thread for 11 mm (snap ring), pkg of 100 ea
Expand
View Pricing
SU860093
Closures for Snap Ring vials,
clear polyethylene cap, red PTFE/silicone, thread for 11 mm (snap ring), pkg of 100 ea
Expand
View Pricing
27426
Closures for Snap Ring vials,
polypropylene seal, PTFE/silicone (with slit), thread for 11 mm snap ring, pkg of 100 ea
Expand
View Pricing
SU860090
Closures for Snap Ring vials,
clear polyethylene cap, PTFE/red rubber, thread for 11 mm snap ring, pkg of 100 ea
Expand
View Pricing
24756
Closures for Snap Ring vials,
polypropylene seal, PTFE/red rubber, diam. 11 mm, pkg of 100 ea
Expand
View Pricing
24754
Closures for Snap Ring vials,
polypropylene seal, PTFE liner, diam. 11 mm, pkg of 100 ea
Expand
View Pricing
24758
Closures for Snap Ring vials,
polypropylene seal, PTFE/silicone, diam. 11 mm, pkg of 100 ea
Expand
View Pricing
SU860113
Closures for Snap Ring vials,
cap size 11 mm, clear polyethylene (with 6.0 mm hole), blue PTFE/white silicone (with slit)
Expand
View Pricing