
Citrate Concentrated Solution

Empirical Formula (Hill Notation):
C6H5Na3O7
CAS Number:
Molecular Weight:
258.07
Products
Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Quality Segment
Sterilization Method
Showing 1-2 of 2