![Chromoionophore III Selectophore™](/deepweb/assets/sigmaaldrich/product/structures/180/118/492a9720-fcc4-44b4-86c2-1e009af913df/640/492a9720-fcc4-44b4-86c2-1e009af913df.png)
Chromoionophore III
![Chromoionophore III Selectophore™](/deepweb/assets/sigmaaldrich/product/structures/180/118/492a9720-fcc4-44b4-86c2-1e009af913df/640/492a9720-fcc4-44b4-86c2-1e009af913df.png)
Synonym(s):
9-(Diethylamino)-5-[(2-octyldecyl)imino]benzo[a]phenoxazine, ETH 5350
Empirical Formula (Hill Notation):
C38H55N3O
CAS Number:
Molecular Weight:
569.86
Products
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing