Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cholera Toxin B subunit
Synonym(s):
CTB
Products
Product Category
bioactive small molecules (5)
stains (1)
Brand
Sigma-Aldrich (5)
Physical Form
Purity
Recombinant Host
Research Area
Chemical Composition
Conjugate
Quality Segment
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
C9903
Cholera Toxin B subunit,
≥95% (SDS-PAGE), lyophilized powder, enterotoxin
Expand
View Pricing
C1655
Cholera Toxin B subunit,
FITC conjugate, lyophilized powder
Expand
View Pricing
SAE0069
Cholera Toxin B subunit,
≥95% (SDS-PAGE), lyophilized powder, enterotoxin
Expand
View Pricing
C9972
Cholera Toxin B subunit,
biotin conjugate, lyophilized powder
Expand
View Pricing
C3741
Cholera Toxin B subunit,
peroxidase conjugate (Contains ~ 2 moles HRP/mole of CTB. ~100 μg HRP conjugated to ~45 μg CTB), lyophilized powder
Expand
View Pricing