Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cesium nitrate
Synonym(s):
Nitric acid cesium salt
Empirical Formula (Hill Notation):
CsNO
3
CAS Number:
7789-18-6
Molecular Weight:
194.91
EC Number:
232-146-8
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (3)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
289337
Cesium nitrate,
99%
Expand
View Pricing
202150
Cesium nitrate,
99.99% trace metals basis
Expand
View Pricing
203041
Cesium nitrate,
99.999% trace metals basis
Expand
View Pricing
562521
Cesium nitrate,
Cabot high-purity grade
Expand
View Pricing