Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
CellCrown
™
inserts
Products
Product Category
plates (7)
inserts (4)
Color
Manufacturer
Wells
Color
Package Size
Pore size (µm)
Sterility
Sterilization Method
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z682004
CellCrown
™
inserts,
96 well plate inserts, 8-well strips, sterile
Expand
View Pricing
Z681806
CellCrown
™
inserts,
6 well plate inserts, sterile
Expand
View Pricing
Z681962
CellCrown
™
inserts,
48 well plate inserts, 6-well strips, sterile
Expand
View Pricing
Z742381
CellCrown
™
inserts,
24 well plate inserts, sterile
Expand
View Pricing
Z681989
CellCrown
™
inserts,
48 well plate inserts with 1 μm PET filter, 6-well strips, sterile
Expand
View Pricing
Z742380
CellCrown
™
inserts,
24 well plate inserts, non-sterile
Expand
View Pricing
Z681997
CellCrown
™
inserts,
96 well plate inserts, 8-well strips, non-sterile
Expand
View Pricing
Z742383
CellCrown
™
inserts,
12 well plate inserts, sterile
Expand
View Pricing
Z681822
CellCrown
™
inserts,
6 well plate inserts with 1.0 μm polycarbonate filter, sterile
Expand
View Pricing
Z681792
CellCrown
™
inserts,
6 well plate inserts, non-sterile
Expand
View Pricing
Z742382
CellCrown
™
inserts,
12 well plate inserts, non-sterile
Expand
View Pricing