
Ceftazidime

Synonym(s):
Ceftazidime pentahydrate
Empirical Formula (Hill Notation):
C22H22N6O7S2 · 5H2O
CAS Number:
Molecular Weight:
636.65
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing