Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cardboard Slide Tray
Products
Product Category
trays (6)
Color
Manufacturer
Height (mm)
Color
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
HS9930
Cardboard Slide Tray,
Holds 30 x slides, black, trim
Expand
View Pricing
HS9914
Cardboard Slide Tray,
Holds 20 x slides, green, trim
Expand
View Pricing
HS9902
Cardboard Slide Tray,
Holds 20 x slides 76x26 mm, black (trim)
Expand
View Pricing
HS9913
Cardboard Slide Tray,
Holds 20 x slides, blue, trim
Expand
View Pricing
HS120196
Cardboard Slide Tray,
Holds 20 x slides 76x26 mm, red (trim)
Expand
View Pricing
HS120197
Cardboard Slide Tray,
Holds 20 x slides 76x26 mm, yellow (trim)
Expand
View Pricing