Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon - Vitreous
Synonym(s):
Carbon - Vitreous
Linear Formula:
C
CAS Number:
7440-44-0
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Element
Filter Products
Showing 1-20 of 28
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF05962765
Carbon - Vitreous,
rod, 100mm, diameter 5.0mm, glassy carbon
Expand
View Pricing
GF07378476
Carbon - Vitreous,
foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm
3
, porosity 96.5%
Expand
View Pricing
GF10355958
Carbon - Vitreous,
foil, 25x25mm, thickness 0.5mm, glassy carbon
Expand
View Pricing
GF20176680
Carbon - Vitreous,
rod, 5mm, diameter 3.0mm, glassy carbon
Expand
View Pricing
GF61342220
Carbon - Vitreous,
foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Expand
View Pricing
GF65427587
Carbon - Vitreous,
rod, 100mm, diameter 3.0mm, glassy carbon
Expand
View Pricing
GF59400233
Carbon - Vitreous,
tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Expand
View Pricing
GF96762864
Carbon - Vitreous,
foil, 50x50mm, thickness 1.0mm, glassy carbon
Expand
View Pricing
GF39088648
Carbon - Vitreous,
tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Expand
View Pricing
GF23723423
Carbon - Vitreous,
rod, 200mm, diameter 1.0mm, glassy carbon
Expand
View Pricing
GF04031132
Carbon - Vitreous,
foam, 300x300mm, thickness 20mm, bulk density 0.05g/cm
3
, porosity 96.5%
Expand
View Pricing
GF22691426
Carbon - Vitreous,
foil, 10x10mm, thickness 1.0mm, glassy carbon
Expand
View Pricing
GF39873369
Carbon - Vitreous,
foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm
3
, porosity 96.5%
Expand
View Pricing
GF93251011
Carbon - Vitreous,
rod, 50mm, diameter 1.0mm, glassy carbon
Expand
View Pricing
GF99431523
Carbon - Vitreous,
foil, 10x10mm, thickness 4.0mm, glassy carbon
Expand
View Pricing
GF45345562
Carbon - Vitreous,
rod, 200mm, diameter 5.0mm, glassy carbon
Expand
View Pricing
GF68195403
Carbon - Vitreous,
rod, 100mm, diameter 7.0mm, glassy carbon
Expand
View Pricing
GF91785134
Carbon - Vitreous,
rod, 200mm, diameter 10mm, glassy carbon
Expand
View Pricing
GF89520521
Carbon - Vitreous,
foil, 25x25mm, thickness 4.0mm, glassy carbon
Expand
View Pricing
GF78447707
Carbon - Vitreous,
rod, 200mm, diameter 3.0mm, glassy carbon
Expand
View Pricing
Page 1 of 2
Page 1 of 2