Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
CAPS
Synonym(s):
3-(Cyclohexylamino)-1-propanesulfonic acid, CAPS, ULTROL
®
Grade
Linear Formula:
C
6
H
11
NH(CH
2
)
3
SO
3
H
CAS Number:
1135-40-6
Molecular Weight:
221.32
EC Number:
214-492-1
Products
Product Category
buffers (3)
Brand
Millipore (1)
Sigma-Aldrich (3)
Color
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Special Grade
Anhydrous (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
C2632
CAPS,
≥99%
Expand
View Pricing
C6070
CAPS,
BioXtra, ≥99%
Expand
View Pricing
239782
CAPS,
ULTROL
®
Grade, ≥99% (HPLC), zwitterionic buffer useful in pH range 7.9-11.1
Expand
View Pricing
RDD015
CAPS,
anhydrous, free-flowing, Redi-Dri
™
, ≥99%
Expand
View Pricing