
Calcium D-gluconate monohydrate

Synonym(s):
D-Gluconic acid calcium salt
Empirical Formula (Hill Notation):
C12H22CaO14 · H2O
CAS Number:
Molecular Weight:
448.39
Products
Product Category
Biological Source
Color
Feature
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing