Skip to Content
Merck
Cadmium foil, light tested, 100x100mm, thickness 0.025mm, 99.7+%

Cadmium

Cadmium foil, light tested, 100x100mm, thickness 0.025mm, 99.7+%
Synonym(s):
Cadmium
Empirical Formula (Hill Notation):
Cd
CAS Number:
Molecular Weight:
112.41

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (m)
Element
Quality Segment
Showing 1-20 of 201
Page 1 of 11
Compare
Product Number
Description
Pricing
Page 1 of 11