![Butylhydroxytoluene European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/427/620/ae45d9a0-d2e5-437b-9020-7adc7a027bad/640/ae45d9a0-d2e5-437b-9020-7adc7a027bad.png)
Butylhydroxytoluene
![Butylhydroxytoluene European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/427/620/ae45d9a0-d2e5-437b-9020-7adc7a027bad/640/ae45d9a0-d2e5-437b-9020-7adc7a027bad.png)
Synonym(s):
Butylhydroxytoluene, 2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, DBPC
Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing