Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bromoethane
Synonym(s):
Ethyl bromide, Bromoethane
Empirical Formula (Hill Notation):
C
2
H
5
Br
CAS Number:
74-96-4
Molecular Weight:
108.97
EC Number:
200-825-8
Products
Brand
Sigma-Aldrich (3)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
124052
Bromoethane,
reagent grade, 98%
Expand
View Pricing
8.00871
Bromoethane,
for synthesis
Expand
View Pricing
239607
Bromoethane,
ReagentPlus
®
, ≥99%
Expand
View Pricing