
Brefeldin A

Synonym(s):
γ,4-Dihydroxy-2-(6-hydroxy-1-heptenyl)-4-cyclopentanecrotonic acid λ-lactone, Ascotoxin, BFA, Cyanein, Decumbin
Empirical Formula (Hill Notation):
C16H24O4
CAS Number:
Molecular Weight:
280.36
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-3 of 3
Compare
Product Number
Description
Pricing