Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Brass alloy, Cu63Zn37
Products
Brand
Sigma-Aldrich (1)
Manufacturer
Physical Form
Diameter (mm)
ID (mm)
Length (m)
Length (mm)
OD (mm)
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
GF07175741
Brass alloy, Cu63Zn37,
foil, 0.5m coil, 0.01mm thickness, hard
Expand
View Pricing
GF88948221
Brass alloy, Cu63Zn37,
1000mm tube, 3.0mm outside diameter, 0.3mm wall thickness, 2.4mm inside diameter, as drawn
Expand
View Pricing
GF16565823
Brass alloy, Cu63Zn37,
1000mm tube, 1.83mm outside diameter, 0.27mm wall thickness, 1.29mm inside diameter, as drawn
Expand
View Pricing
GF25606368
Brass alloy, Cu63Zn37,
200mm tube, 1.5mm outside diameter, 0.25mm wall thickness, 1mm inside diameter, as drawn
Expand
View Pricing
GF03835593
Brass alloy, Cu63Zn37,
1000mm tube, 3.0mm outside diameter, 0.5mm wall thickness, 2mm inside diameter, as drawn
Expand
View Pricing
GF93298269
Brass alloy, Cu63Zn37,
200mm tube, 2.26mm outside diameter, 0.28mm wall thickness, 1.7mm inside diameter, as drawn
Expand
View Pricing
GF28998938
Brass alloy, Cu63Zn37,
200mm tube, 1.18mm outside diameter, 0.23mm wall thickness, 0.72mm inside diameter, as drawn
Expand
View Pricing
GF09010932
Brass alloy, Cu63Zn37,
100mm rod, 5.0mm diameter, as drawn
Expand
View Pricing
GF62283630
Brass alloy, Cu63Zn37,
500mm tube, 2.26mm outside diameter, 0.28mm wall thickness, 1.7mm inside diameter, as drawn
Expand
View Pricing
GF72709364
Brass alloy, Cu63Zn37,
31mm tube, 1.18mm outside diameter, 0.23mm wall thickness, 0.72mm inside diameter, as drawn
Expand
View Pricing
GF81901740
Brass alloy, Cu63Zn37,
foil, 100 x 100mm, 0.05mm thickness, half hard, light tight (lt)
Expand
View Pricing
GF53606875
Brass alloy, Cu63Zn37,
foil, 150 x 150mm, 0.025mm thickness, hard, light tight (lt)
Expand
View Pricing
GF35390066
Brass alloy, Cu63Zn37,
100mm tube, 1.83mm outside diameter, 0.27mm wall thickness, 1.29mm inside diameter, as drawn
Expand
View Pricing
GF67694653
Brass alloy, Cu63Zn37,
1000mm tube, 0.76mm outside diameter, 0.25mm wall thickness, 0.26mm inside diameter, as drawn
Expand
View Pricing
GF32541555
Brass alloy, Cu63Zn37,
500mm tube, 0.76mm outside diameter, 0.25mm wall thickness, 0.26mm inside diameter, as drawn
Expand
View Pricing
GF86608660
Brass alloy, Cu63Zn37,
foil, 25 x 25mm, 0.008mm thickness, hard, light tight (lt)
Expand
View Pricing
GF28859248
Brass alloy, Cu63Zn37,
500mm rod, 10.0mm diameter, annealed
Expand
View Pricing
GF95839936
Brass alloy, Cu63Zn37,
500mm rod, 5.0mm diameter, as drawn
Expand
View Pricing
GF07757376
Brass alloy, Cu63Zn37,
500mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
GF23494899
Brass alloy, Cu63Zn37,
100mm rod, 2.0mm diameter, as drawn
Expand
View Pricing
Page 1 of 11
Page 1 of 11