Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Brass
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
OD (mm)
Particle Size (µm)
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
GF14336276
Brass,
powder, Cu 70%/Zn 30%, max. particle size 100 micron, 2_kg, condition atomized
Expand
View Pricing
GF21550890
Brass,
foil, Cu 63%/Zn 37%, thickness 1.42 mm, size 25 × 25 mm, temper half hard
Expand
View Pricing
GF72955301
Brass,
wire, Cu63%/Zn37%, length 100 m, 0.08 mm diameter, temper hard
Expand
View Pricing
GF64464060
Brass,
tube, outside diameter 3.17 mm, Cu63%/Zn37%, length 1000 mm, wall thickness 0.455 mm
Expand
View Pricing
GF94915716
Brass,
tube, Cu63%/Zn37%, length 500 mm, outside diameter 3.17 mm, wall thickness 0.455 mm
Expand
View Pricing
GF15613927
Brass,
powder, Cu 70%/Zn 30%, max. particle size 100 micron, 0.1_kg, condition atomized
Expand
View Pricing
GF98032293
Brass,
powder, 100 max. part. size (micron), Cu70%/Zn30%, weight 0.2 kg, condition atomized
Expand
View Pricing
GF40269270
Brass,
powder, Cu 70%/Zn 30%, max. particle size 100 micron, 1_kg, condition atomized
Expand
View Pricing
GF64417495
Brass,
wire, Cu63%/Zn37%, 0.08 mm diameter, length 250 m, temper hard
Expand
View Pricing
GF31159324
Brass,
tube, Cu 63%/Zn 37%, outside diameter 3.17 mm, length 200 mm, wall thickness 0.455 mm
Expand
View Pricing
GF35377713
Brass,
wire, Cu 63%/Zn 37%, 0.08 mm diameter, length 1000 m, temper hard
Expand
View Pricing
GF48335014
Brass,
wire, Cu 63%/Zn 37%, 0.08 mm diameter, length 5000 m, temper hard
Expand
View Pricing
GF75183611
Brass,
wire, Cu63%/Zn37%, 0.08 mm diameter, length 500 m, temper hard
Expand
View Pricing
GF72832192
Brass,
wire, 0.08 mm diameter, Cu63%/Zn37%, length 2000 m, temper hard
Expand
View Pricing
GF13963653
Brass,
powder, Cu 70%/Zn 30%, max. particle size 100 micron, 0.5_kg, condition atomized
Expand
View Pricing
GF79633569
Brass,
foil, Cu63%/Zn37%, thickness 0.5 mm, length 0.5 m, temper half hard
Expand
View Pricing
GF79449272
Brass,
tube, Cu63%/Zn37%, outside diameter 3.17 mm, length 100 mm, wall thickness 0.455 mm
Expand
View Pricing