Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
volumetric flask BLAUBRAND
®
USP
Products
Product Category
flasks (20)
Manufacturer
Capacity (mL)
Glassware Class
Color
Package Size
Filter Products
Showing 1-20 of 20
Compare
Product Number
Description
Pricing
BR37465
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 50 mL, joint: 12/21, glass stopper
Expand
View Pricing
BR37464
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 25 mL, joint: 10/19, glass stopper
Expand
View Pricing
BR37467
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 100 mL, joint: 14/23, glass stopper
Expand
View Pricing
BR37462
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 10 mL, neck joint: ST/NS 10/19, glass stopper
Expand
View Pricing
BR37488
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 200 mL, joint: 14/23, polypropylene stopper
Expand
View Pricing
BR37490
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 500 mL, joint: 19/26, polypropylene stopper
Expand
View Pricing
BR37491
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 1000 mL, joint: 24/29, polypropylene stopper
Expand
View Pricing
BR37463
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 20 mL, joint: 10/19, glass stopper
Expand
View Pricing
BR37470
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 500 mL, joint: 19/26, glass stopper
Expand
View Pricing
BR37487
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 100 mL, joint: 14/23, polypropylene stopper
Expand
View Pricing
BR37482
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 10 mL, joint: 10/19, polypropylene stopper
Expand
View Pricing
BR37471
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 1000 mL, joint: 24/29, glass stopper
Expand
View Pricing
BR37468
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 200 mL, joint: 14/23, glass stopper
Expand
View Pricing
BR37469
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 250 mL, joint: 14/23, glass stopper
Expand
View Pricing
BR37483
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 20 mL, joint: 10/19, polypropylene stopper
Expand
View Pricing
BR37489
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 250 mL, joint: 14/23, polypropylene stopper
Expand
View Pricing
BR37461
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 5 mL, neck joint: ST/NS 10/19, glass stopper
Expand
View Pricing
BR37485
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 50 mL, joint: 12/21, polypropylene stopper
Expand
View Pricing
BR37484
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 25 mL, joint: 10/19, polypropylene stopper
Expand
View Pricing
BR37481
BRAND
®
volumetric flask BLAUBRAND
®
USP,
capacity 5 mL, joint: 10/19, polypropylene stopper
Expand
View Pricing