Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
tray stackable
Products
Product Category
trays (6)
Manufacturer
Color
Package Size
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BR156642
BRAND
®
tray stackable,
size 300 x 240 x 70 mm
Expand
View Pricing
BR156640
BRAND
®
tray stackable,
size 225 x 180 x 45 mm
Expand
View Pricing
BR156646
BRAND
®
tray stackable,
size 510 x 410 x 120 mm
Expand
View Pricing
BR156644
BRAND
®
tray stackable,
size 370 x 310 x 75 mm
Expand
View Pricing
BR156648
BRAND
®
tray stackable,
size 625 x 530 x 140 mm
Expand
View Pricing
BR156650
BRAND
®
tray stackable,
size 840 x 645 x 160 mm
Expand
View Pricing