Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
Transferpettor seals
Products
Manufacturer
Color
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z328588
BRAND
®
Transferpettor seals,
for 50 μL caps, yellow PTFE
Expand
View Pricing
Z328626
BRAND
®
Transferpettor seals,
for 200-1000 μL caps, clear PE
Expand
View Pricing
Z328634
BRAND
®
Transferpettor seals,
for 1-5 mL caps, clear PE
Expand
View Pricing
Z328642
BRAND
®
Transferpettor seals,
for 2-10 mL caps, clear PE
Expand
View Pricing
Z328596
BRAND
®
Transferpettor seals,
for 100-200 μL caps, yellow PTFE
Expand
View Pricing
Z328561
BRAND
®
Transferpettor seals,
for 20-25 μL caps, yellow PTFE
Expand
View Pricing
Z328618
BRAND
®
Transferpettor seals,
for 100-500 μL caps, clear PE
Expand
View Pricing