Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
Transferpettor positive displacement pipette, digital volume
Products
Product Category
pipettors (8)
Manufacturer
Volume (ml)
Volume (µL)
Electronic
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z328413
BRAND
®
Transferpettor positive displacement pipette, digital volume,
volume 1-5 mL, accuracy: 0.5%, precision: 0.2%
Expand
View Pricing
Z328421
BRAND
®
Transferpettor positive displacement pipette, digital volume,
volume 2-10 mL, accuracy: 0.5%, precision: 0.2%
Expand
View Pricing
Z328405
BRAND
®
Transferpettor positive displacement pipette, digital volume,
volume 200-1000 μL, accuracy: 0.5%, precision: 0.2%
Expand
View Pricing
Z328391
BRAND
®
Transferpettor positive displacement pipette, digital volume,
volume 100-500 μL, accuracy: 0.5%, precision: 0.2%
Expand
View Pricing
Z328383
BRAND
®
Transferpettor positive displacement pipette, digital volume,
volume 20-100 μL, accuracy: 1.5%, precision: 0.4%
Expand
View Pricing
Z328359
BRAND
®
Transferpettor positive displacement pipette, digital volume,
volume 2.5-10 μL, accuracy: 3.0%, precision: 0.8%
Expand
View Pricing
Z328375
BRAND
®
Transferpettor positive displacement pipette, digital volume,
volume 10-50 μL, accuracy: 1.8%, precision: 0.4%
Expand
View Pricing
Z328367
BRAND
®
Transferpettor positive displacement pipette, digital volume,
volume 5-25 μL, accuracy: 2.4%, precision: 0.5%
Expand
View Pricing