Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
sealing film for microplates
Products
Product Category
sealing films and foils (10)
Feature
Manufacturer
Color
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
BR701365
BRAND
®
sealing film for microplates,
self-adhesive, gas permeable, suitable for (for cell and tissue culture)
Expand
View Pricing
BR781390
BRAND
®
sealing film for microplates,
self-adhesive, suitable for (PCR), polypropylene
Expand
View Pricing
BR701364
BRAND
®
sealing film for microplates,
self-adhesive, gas permeable, suitable for (for cell and tissue culture)
Expand
View Pricing
BR701372
BRAND
®
sealing film for microplates,
self-adhesive, suitable for (for luminescence measurement), vinyl
Expand
View Pricing
BR701371
BRAND
®
sealing film for microplates,
self-adhesive, suitable for (for storage/fluorescence measurement), vinyl
Expand
View Pricing
BR781381
BRAND
®
sealing film for microplates,
self-adhesive, suitable for (for cold storage), aluminum
Expand
View Pricing
BR701370
BRAND
®
sealing film for microplates,
self-adhesive, suitable for (for automation), polypropylene
Expand
View Pricing
BR701374
BRAND
®
sealing film for microplates,
self-adhesive, suitable for storage, vinyl (with pre-pierced zone)
Expand
View Pricing
BR701377
BRAND
®
sealing film for microplates,
suitable for PCR, storage, aluminum film, DMSO/solvent resistant, operating temperature (-80)—120 °C, pack of 100 ea
Expand
View Pricing
BR701376
BRAND
®
sealing film for microplates,
suitable for PCR, storage, PET film, DMSO/solvent resistant, operating temperature (-80)—120 °C, pack of 100 ea
Expand
View Pricing