Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
PP microtube, attached screw cap with silicone seal
Products
Product Category
tubes (5)
Manufacturer
Capacity (mL)
Diameter (mm)
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
Z637416
BRAND
®
PP microtube, attached screw cap with silicone seal,
capacity 1.5 mL, graduations, up to 1 mL, self-standing bottom
Expand
View Pricing
Z637432
BRAND
®
PP microtube, attached screw cap with silicone seal,
capacity 2.0 mL, graduations, up to 1.2 mL, self-standing bottom
Expand
View Pricing
Z637408
BRAND
®
PP microtube, attached screw cap with silicone seal,
capacity 0.5 mL, graduations: no, self-standing bottom
Expand
View Pricing
Z637424
BRAND
®
PP microtube, attached screw cap with silicone seal,
capacity 1.5 mL, graduations, up to 1 mL, conical bottom
Expand
View Pricing
Z637440
BRAND
®
PP microtube, attached screw cap with silicone seal,
capacity 2.0 mL, graduations, up to 1.4 mL, round bottom
Expand
View Pricing