Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
PP graduated centrifuge tube, screw cap
Products
Product Category
tubes (6)
Manufacturer
Volume (ml)
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BR114822
BRAND
®
PP graduated centrifuge tube, screw cap,
volume 50 mL, with base, non-sterile
Expand
View Pricing
BR114821
BRAND
®
PP graduated centrifuge tube, screw cap,
volume 50 mL, without base, sterile
Expand
View Pricing
BR114823
BRAND
®
PP graduated centrifuge tube, screw cap,
volume 50 mL, with base, sterile
Expand
View Pricing
BR114818
BRAND
®
PP graduated centrifuge tube, screw cap,
volume 15 mL, without base, sterile
Expand
View Pricing
BR114820
BRAND
®
PP graduated centrifuge tube, screw cap,
volume 50 mL, without base, non-sterile
Expand
View Pricing
BR114817
BRAND
®
PP graduated centrifuge tube, screw cap,
volume 15 mL, without base, non-sterile
Expand
View Pricing