Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
pipette tips, racked, TipRack
Products
Product Category
pipette tips (8)
Manufacturer
Volume (µL)
Color
Package Size
Sterility
Sterilization Method
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z740120
BRAND
®
pipette tips, racked, TipRack,
volume 50-1000 μL, non-sterile
Expand
View Pricing
Z740118
BRAND
®
pipette tips, racked, TipRack,
volume 2-200 μL, non-sterile
Expand
View Pricing
Z740112
BRAND
®
pipette tips, racked, TipRack,
volume 0.1-20 μL, non-sterile
Expand
View Pricing
Z740114
BRAND
®
pipette tips, racked, TipRack,
volume 0.5-20 μL, non-sterile
Expand
View Pricing
Z740117
BRAND
®
pipette tips, racked, TipRack,
volume 2-200 μL, BIO-CERT
™
Expand
View Pricing
Z740122
BRAND
®
pipette tips, racked, TipRack,
volume 5-300 μL, non-sterile
Expand
View Pricing
Z740113
BRAND
®
pipette tips, racked, TipRack,
volume 0.5-20 μL, BIO-CERT
®
Expand
View Pricing
Z740121
BRAND
®
pipette tips, racked, TipRack,
volume 5-300 μL, BIO-CERT
®
Expand
View Pricing