Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
pipette tips, bulk
Products
Product Category
pipette tips (10)
Manufacturer
Volume (µL)
Color
Package Size
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z740030
BRAND
®
pipette tips, bulk,
volume 2-200 μL, pack of 1000 ea (1 bag of 1000)
Expand
View Pricing
Z740031
BRAND
®
pipette tips, bulk,
volume 50-1000 μL, pack of 1000 ea (2 bags of 500)
Expand
View Pricing
Z740095
BRAND
®
pipette tips, bulk,
volume 50-1000 μL, pack of 5000 ea (10 bags of 500)
Expand
View Pricing
Z740091
BRAND
®
pipette tips, bulk,
volume 0.1-20 μL, pack of 2000 ea (2 bags of 1000)
Expand
View Pricing
Z740094
BRAND
®
pipette tips, bulk,
volume 2-200 μL, pack of 10,000 ea (10 bags of 1000)
Expand
View Pricing
Z740032
BRAND
®
pipette tips, bulk,
volume 5-300 μL, pack of 1000 ea (1 bag of 1000)
Expand
View Pricing
Z740092
BRAND
®
pipette tips, bulk,
volume 0.5-20 μL, pack of 10,000 ea (10 bags of 1000)
Expand
View Pricing
Z740028
BRAND
®
pipette tips, bulk,
volume 0.5-20 μL, pack of 2000 ea (2 bags of 1000)
Expand
View Pricing
Z740027
BRAND
®
pipette tips, bulk,
volume 0.1-20 μL, pack of 10,000 ea (10 bags of 1000)
Expand
View Pricing
Z740096
BRAND
®
pipette tips, bulk,
volume 5-300 μL, pack of 10,000 ea (10 bags of 1000)
Expand
View Pricing