Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
graduated cylinder, PMP
Products
Product Category
cylinders (8)
Brand
Sigma-Aldrich (1)
Manufacturer
Height (mm)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z327476
BRAND
®
graduated cylinder, PMP,
volume 1,000 mL, tol. accuracy: 5 mL, subdivision, 10 mL
Expand
View Pricing
Z327263
BRAND
®
graduated cylinder, PMP,
volume 10 mL, tol. accuracy: 0.08 mL, subdivision, 0.2 mL
Expand
View Pricing
Z327379
BRAND
®
graduated cylinder, PMP,
volume 100 mL, tol. accuracy: 0.4 mL, subdivision, 1 mL
Expand
View Pricing
Z327409
BRAND
®
graduated cylinder, PMP,
volume 250 mL, tol. accuracy: 0.8 mL, subdivision, 2 mL
Expand
View Pricing
Z327344
BRAND
®
graduated cylinder, PMP,
volume 50 mL, tol. accuracy: 0.4 mL, subdivision, 1 mL
Expand
View Pricing
Z327301
BRAND
®
graduated cylinder, PMP,
volume 25 mL, tol. accuracy: 0.20 mL, subdivision, 0.5 mL
Expand
View Pricing
Z327433
BRAND
®
graduated cylinder, PMP,
volume 500 mL, tol. accuracy: 2.0 mL, subdivision, 5 mL
Expand
View Pricing
Z327506
BRAND
®
graduated cylinder, PMP,
volume 2,000 mL, tol. accuracy: 8 mL, subdivision, 20 mL
Expand
View Pricing