Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
glass bottle-top tilt pipette dispenser
Products
Product Category
dispensers (6)
Color
Manufacturer
Volume (ml)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BR43078
BRAND
®
glass bottle-top tilt pipette dispenser,
without bottle
Expand
View Pricing
BR43055
BRAND
®
glass bottle-top tilt pipette dispenser,
without bottle
Expand
View Pricing
BR43058
BRAND
®
glass bottle-top tilt pipette dispenser,
without bottle
Expand
View Pricing
BR43070
BRAND
®
glass bottle-top tilt pipette dispenser,
without bottle
Expand
View Pricing
BR43088
BRAND
®
glass bottle-top tilt pipette dispenser,
without bottle
Expand
View Pricing
BR43066
BRAND
®
glass bottle-top tilt pipette dispenser,
without bottle
Expand
View Pricing