Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass
Products
Product Category
flasks (11)
Brand
Sigma-Aldrich (1)
Manufacturer
Capacity (mL)
Glassware Class
Package Size
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
Z326445
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass,
volume 100 mL, accuracy: 0.1 mL
Expand
View Pricing
Z326437
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass,
volume 50 mL, accuracy: 0.08 mL
Expand
View Pricing
Z326399
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass,
volume 10 mL, accuracy: 0.04 mL
Expand
View Pricing
Z326402
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass,
volume 20 mL, accuracy: 0.04 mL
Expand
View Pricing
Z326488
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass,
volume 500 mL, accuracy: 0.25 mL
Expand
View Pricing
Z326380
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass,
volume 5 mL, accuracy: 0.04 mL
Expand
View Pricing
Z326410
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass,
volume 25 mL, accuracy: 0.04 mL
Expand
View Pricing
Z326496
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass,
volume 1000 mL, accuracy: 0.4 mL
Expand
View Pricing
Z326453
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass,
volume 200 mL, accuracy: 0.15 mL
Expand
View Pricing
Z326429
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass,
volume 50 mL, accuracy: 0.06 mL
Expand
View Pricing
Z326461
BRAND
®
BLAUBRAND
®
volumetric flask, PP stopper, amber glass,
volume 250 mL, accuracy: 0.15 mL
Expand
View Pricing